Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACSM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549396

Catalog No. PA549396

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
Specifications

Specifications

ACSM5
Polyclonal
PBS with 0.09% sodium azide
Q6NUN0, Q8BGA8, Q6AYT9
Acyl-coenzyme A synthetase ACSM5, mitochondrial, Acyl-CoA synthetase medium-chain family member 5, ACSM5, MACS3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
272428, 361637, 54988
Western Blot
Unconjugated
ACSM5
Liquid
ACSM5
KLH conjugated synthetic peptide between 562-596 amino acids from the C-terminal region of human ACSM5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit