Learn More
Abnova™ ACSS2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00055902T01
Catalog No. 89-932-418
Add to cart
Description
Description
Specifications
Specifications
293T
NM_139274.1
pCMV-ACSS2 full-length
100 μL
Human
ACSS2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 66.77kDa
Transient overexpression cell lysate was tested with anti-ACSS2 antibody (Mfr. No. H00055902-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot