Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTH Rabbit anti-Human, Clone: CLIP/2859R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen ACTH
Clone CLIP/2859R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122740
View Documents
Novus Biologicals
NBP30763320UG
20 μg
Each for $159.82
Add to cart
 
NB122739
View Documents
Novus Biologicals
NBP30763310UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

ACTH Monoclonal antibody specifically detects ACTH in Human samples. It is validated for ELISA
Specifications

Specifications

ACTH
0.2 mg/ml
ELISA
Purified
RUO
Human
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF)
Monoclonal
Protein A or G purified
CLIP/2859R
ELISA
Unconjugated
Rabbit
Neuroscience
10 mM PBS with 0.05% BSA
5443
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit