Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACTH Rabbit anti-Human, Mouse, Rat, Clone: CLIP/2040R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP30762720UG

 View more versions of this product

Catalog No. NB122728


Add to Cart

Description

Description

ACTH Monoclonal specifically detects ACTH in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

ACTH
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Synthetic peptide corresponding to aa1-24 of human ACTH
20 μg
Neuroscience
5443
Store at 4°C.
IgG
Immunohistochemistry (Paraffin)
CLIP/2040R
Unconjugated
10 mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit