Learn More
HTL-Strefa Acti-Lance Baby Safety Heelstick
Sterile, single-use safety lancets are push-button activated medical devices used to perform finger pricks to collect micro samples of capillary blood.
Manufacturer:  HTL-Strefa 8714
 View more versions of this product
Catalog No. 22-192-018
Add to cart
Description
Description
  • Fully enclosed needle housing to prevent sharps injuries
  • Long proctective cap for easy removal
  • Protective cap for needle safety
  • T-shape for firm and intuitive grip
  • Wide lancet end enables precise and convenient positioning on the test site
Specifications
Specifications
Yellow
2.20 mm
50ct. Per box
Safety Lancet
Micropreemie blood testing
Medical Sharp
0.60 mm