Learn More
HTL-Strefa Acti-Lance Baby Safety Heelstick
Sterile, single-use safety lancets are push-button activated medical devices used to perform finger pricks to collect micro samples of capillary blood.
Catalog Number | Mfr. No. | For Use With (Application) | Color | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Color | Size | Price | Quantity & Availability | |||
22-192-019
|
HTL-Strefa
8715 |
Preemie blood testing | Pink | 0.85 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
22-192-020
|
HTL-Strefa
8716 |
Baby heelstick blood testing | Blue | 1.00 mm | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Fully enclosed needle housing to prevent sharps injuries
- Long proctective cap for easy removal
- Protective cap for needle safety
- T-shape for firm and intuitive grip
- Wide lancet end enables precise and convenient positioning on the test site
Specifications
Safety Lancet | |
Medical Sharp |