Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin Mouse anti-Human, Mouse, Rat, Porcine, Bovine, Equine, PE-Cyanine7, Clone: 5J11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP225142PECY7
Description
Actin Monoclonal antibody specifically detects Actin in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Flow Cytometry.Specifications
Actin | |
5J11 | |
Flow Cytometry | |
Affinity Purified, Conjugated | |
ACTA1 | |
An Actin preparation derived from bovine brain. | |
Immunogen affinity purified | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
Primary | |
58.0 | |
Bovine, Equine, Human, Mouse, Porcine, Rat |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
This antibody recognizes all six isotypes of Actin. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title