Learn More
Actin, Muscle Specific, Ab-6 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.
Supplier: Epredia MS1296P1
Description
Host Species: Mouse
Clone: MSA06; same as HUC1-1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat, Pig, Horse, Cow and Dog. Others not tested.
Epitope: Not determined
Immunogen: Purified actin
Positive Control: Muscle or sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
Actin, Muscle Specific Ab-6 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Muscle Biology | |
Bovine, Canine, Equine, Human, Mouse, Porcine, Rat | |
IgG1 κ |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
MSA06; same as HUC1-1 | |
Unconjugated | |
Purified actin | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.