Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.

$151.00 - $477.50

Specifications

Antigen Actin, Muscle Specific Ab-6
Clone MSA06; same as HUC1-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1296P
View Documents
Thermo Scientific™
MS1296P
Purified Ab with BSA and Azide 1mL
Each for $417.00
Add to cart
 
MS1296P0
View Documents
Thermo Scientific™
MS1296P0
Purified Ab with BSA and Azide 100μL
Each for $151.00
Add to cart
 
MS1296P1
View Documents
Thermo Scientific™
MS1296P1
Purified Ab with BSA and Azide 500μL
Each for $297.00
Add to cart
 
MS1296P1ABX
View Documents
Thermo Scientific™
MS1296P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $377.50
Add to cart
 
MS1296PABX
View Documents
Thermo Scientific™
MS1296PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $477.50
Add to cart
 
MS1296R7
View Documents
Thermo Scientific™
MS1296R7
Ready-to-use for IHC (UVLP) 7mL
Each for $221.50
Add to cart
 
MS1296R1
View Documents
Thermo Scientific™
MS1296R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Muscle Specific Ab-6, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: MSA06; same as HUC1-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat, Pig, Horse, Cow and Dog. Others not tested.

Epitope: Not determined

Immunogen: Purified actin

Positive Control: Muscle or sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Actin, Muscle Specific Ab-6
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG1 kappa
MSA06; same as HUC1-1
Monoclonal
Murine
Muscle Biology
Purified actin
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit