Learn More
Actin (muscle specific) Mouse anti-Human, Rat, Clone: MSA/953, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB14633
Catalog No. 89-034-716
Add to cart
Description
Description
Mouse monoclonal antibody raised against synthetic peptide of human muscle actin.
  • High specificity
  • High reactivity
Specifications
Specifications
Actin (muscle specific)
MSA/953
Mouse monoclonal antibody raised against synthetic peptide of human muscle actin.
In 10 mM PBS (0.05% BSA and 0.05% azide).
Liquid
ACTA1
A synthetic peptide corresponding to human muscle actins.
Protein A/G purification
RUO
Primary
58|59|70
Human, Rat
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determin
P62736; P68032; P68133
ACTA|ASMA|CFTD|CFTD1|CFTDM|MPFD|NEM1|NEM2|NEM3
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
This antibody recognizes actin of skeletal, cardiac, and smooth muscle cells. It is not reactive with other mesenchymal cells except for myoepithelium.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)