Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin (Muscle Specific) Mouse anti-Human, Rat, Rabbit, Clone: rMSA/953, Novus Biologicals™
Mouse Monoclonal Antibody
$208.09 - $439.77
Specifications
Antigen | Actin (Muscle Specific) |
---|---|
Clone | rMSA/953 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 0.5-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Actin (Muscle Specific) | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
10 mM PBS with 0.05% BSA | |
ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
Recombinant protein corresponding to human muscle actin | |
Primary | |
Store at 4°C. |
rMSA/953 | |
Western Blot 0.5-1.0 ug/ml, Immunohistochemistry-Paraffin 0.5-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Rabbit | |
P62736, P68133, P68032 | |
58 | |
IgG1 κ | |
Protein A or G purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title