Learn More
Enzo Life SciencesSupplier Diversity Partner Actin polyclonal antibody (50 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADICSA400D
Host: Rabbit. Immunogen: Bovine skeletal muscle Actin. UniProt: P68133. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol.
Catalog No. 50-659-005