Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Actin, pan Ab-5, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.

Manufacturer:  Thermo Scientific™ MS1295P

 View more versions of this product

Catalog No. MS1295P

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments with Thermo Scientific™ Actin, pan Ab-5, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: ACTN05; same as C4

Isotype: IgG1/κ

Species Reactivity: Human, Cow, Mouse, Dog, Pig, Rabbit, Rat, Chicken, Dictyostelium discoideum and Physarum polycephalum. Others not known.

Epitope: Not determined

Immunogen: Chicken gizzard actin

Positive Control: HeLa cells

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Actin, pan Ab-5
Monoclonal
200μg/mL
Concentrated
Chicken gizzard actin
1mL
Muscle Biology
Primary
Bovine, Canine, Chicken, Human, Murine, Physarum polycephalum, Porcine, Rabbit, Rat
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
ACTN05; same as C4
Unconjugated
Murine
IgG1 kappa
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit