Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ β-Actin Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and Western blotting with Thermo Scientific™ Actin β, Rabbit Polyclonal Antibody.

Specifications

Antigen Actin beta
Clone Polyclonal
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB9421P1
View Documents
Thermo Scientific™
RB9421P1
500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9421P0
View Documents
Thermo Scientific™
RB9421P0
100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9421P
View Documents
Thermo Scientific™
RB9421P
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
RB9421R7
View Documents
Thermo Scientific™
RB9421R7
7mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and Western blotting with Thermo Scientific™ Actin β, Rabbit Polyclonal Antibody.

The two major cytoskeletal proteins implicated in cell motility are actin and myosin. There are three α-actin (skeletal, cardiac, and smooth muscle), one β-actin (β nonmuscle), and two γ-actins (γ-smooth muscle and γ-nonmuscle). Antibody to β-actin provides a specific and useful tool in studying the intracellular distribution of β-actin and the static and dynamic aspects of the cytoskeleton.

Host Species: Rabbit

Species Reactivity: Human, Mouse, Rat, Bovine, Sheep, Pig, Rabbit, Cat, Dog, Guinea Pig, Chicken, Hirudo medicinalis (leech) and Drosophila.

Epitope: N-terminus

Immunogen: Synthetic peptide corresponding to N-terminus of human β-actin

Positive Control: Human leiomyoma

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Actin beta
Western Blot
Unconjugated
RUO
Primary
Polyclonal
Polyclonal
Rabbit
Cytoskeletal Protein
Synthetic peptide corresponding to N-terminus of human beta-actin
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit