Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Actin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin Skeletal Muscle Ab-2 Mouse Monoclonal Antibody.

$151.00 - $446.00

Specifications

Antigen Actin, Skeletal Muscle Ab-2
Clone 5C5.F8.C7; or alpha-Sr-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS185P
View Documents
Thermo Scientific™
MS185P
Purified Ab with BSA and Azide 1mL
Each for $446.00
Add to cart
 
MS185P0
View Documents
Thermo Scientific™
MS185P0
Purified Ab with BSA and Azide 100μL
Each for $151.00
Add to cart
 
MS185P1
View Documents
Thermo Scientific™
MS185P1
Purified Ab with BSA and Azide 500μL
Each for $337.50
Add to cart
 
MS185R7
View Documents
Thermo Scientific™
MS185R7
Ready-to-use for IHC (UVLP) 7mL
Each for $221.50
Add to cart
 
MS185R1
View Documents
Thermo Scientific™
MS185R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 5C5.F8.C7 (α-Sr-1)

Isotype: IgM/κ

Species Reactivity: Human, Cow, Rabbit, Sheep, Rat, Guinea Pig and Frog. Others not known.

Epitope: N-terminal decapeptide

Immunogen: N-Terminal decapeptide of α skeletal muscle isoform of actin; acetylated at the N-terminus

Positive Control: Skeletal or Cardiac muscle, or Rhabdomyosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Actin, Skeletal Muscle Ab-2
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
5C5.F8.C7; or alpha-Sr-1
Monoclonal
Murine
Muscle Biology
IgM kappa
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit