Learn More
Actin, Smooth Muscle, Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Epredia™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.
Supplier: Epredia MS113P1ABX
Description
Host Species: Mouse
Clone: 1A4; also known as asm-1
Isotype: IgG2a/κ
Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.
Epitope: Acetyl group and the first four amino acids on the N-terminal end of the peptidic chain of α-smooth actin
Immunogen: N-terminal decapeptide of α-smooth muscle isoform of actin; acetylated at the N-terminus
Positive Control: Smooth muscle or leiomyosarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
Actin, Smooth Muscle Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Muscle Biology | |
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit, Rat | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1A4 ; also known as asm-1 | |
Unconjugated | |
N-terminal decapeptide of alpha-smooth muscle isoform of actin; acetylated at the N-terminus | |
IVD | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.