Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.

$265.00 - $834.00

Specifications

Antigen Actin, Smooth Muscle Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
Regulatory Status IVD
View More Specs

Products 11
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS113B
View Documents
Thermo Scientific™
MS113B
Biotinylated Ab with BSA and Azide 1mL
Each for $834.00
Add to cart
 
MS113B0
View Documents
Thermo Scientific™
MS113B0
Biotinylated Ab with BSA and Azide 100μL
Each for $265.00
Add to cart
 
MS113B1
View Documents
Thermo Scientific™
MS113B1
Biotinylated Ab with BSA and Azide 500μL
Each for $601.00
Add to cart
 
MS113P
View Documents
Thermo Scientific™
MS113P
Purified Ab with BSA and Azide 1mL
Each for $618.00
Add to cart
 
MS113P1
View Documents
Thermo Scientific™
MS113P1
Purified Ab with BSA and Azide 500μL
Each for $490.00
Add to cart
 
MS113P1ABX
View Documents
Thermo Scientific™
MS113P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $419.50
Add to cart
 
MS113PABX
View Documents
Thermo Scientific™
MS113PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $637.00
Add to cart
 
MS113R7
View Documents
Thermo Scientific™
MS113R7
Ready-to-use for IHC (UVLP) 7mL
Each for $303.00
Add to cart
 
MS113RQ
View Documents
Thermo Scientific™
MS113RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $431.00
Add to cart
 
MS113P0
View Documents
Thermo Scientific™
MS113P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS113R1
View Documents
Thermo Scientific™
MS113R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Actin, Smooth Muscle Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 1A4; also known as asm-1

Isotype: IgG2a/κ

Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not known.

Epitope: Acetyl group and the first four amino acids on the N-terminal end of the peptidic chain of α-smooth actin

Immunogen: N-terminal decapeptide of α-smooth muscle isoform of actin; acetylated at the N-terminus

Positive Control: Smooth muscle or leiomyosarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

<
Specifications

Specifications

Actin, Smooth Muscle Ab-1
Monoclonal
IVD
Primary
Immunofluorescence, Immunohistochemistry (Paraffin)
Murine
Muscle Biology
N-terminal decapeptide of alpha-smooth muscle isoform of actin; acetylated at the N-terminus
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit