Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smooth Muscle Actin Rabbit Polyclonal Antibody, Epredia™

Stain smooth muscle cells in vessel walls, gut wall, and myometrium with Epredia™ Actin, Smooth Muscle, Rabbit Polyclonal Antibody.

Supplier:  Epredia RB9010P1

 View more versions of this product

Catalog No. RB9010P1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Baboon, Cow, Rabbit, Mouse, Rat and Chicken. Others not tested.

Epitope: N-terminus

Immunogen: A synthetic peptide from N-terminus of human α-smooth muscle isoform of actin

Positive Control: Smooth muscle of arteries, gut wall, uterus and Leiomyoma

Cellular Localization: Cytoplasmic.

This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Actin, Smooth Muscle
Polyclonal
Unconjugated
A synthetic peptide from N-terminus of human alpha-smooth muscle isoform of actin
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
200µg/mL
Rabbit
500 μL
Muscle Biology
Bovine, Chicken, Human, Mouse, Non-human Primate, Rabbit
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit