Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin11 Mouse anti-Plant, Alexa Fluor™ 532, Clone: mAb2345a, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP253120AF532

Catalog No. NP23120A32


Add to cart

Description

Description

Actin11 Monoclonal antibody specifically detects Actin11 in Plant samples. It is validated for Western Blot, Immunofluorescence.
Specifications

Specifications

Actin11
mAb2345a
Western Blot, Immunofluorescence
Conjugated, Purified
Purified Arabidopsis actin (ACT11)
Protein G purified
Store at 4°C in the dark.
Monoclonal
Plant
Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
0.1 mL
Primary
This mouse monoclonal antibody was generated against plant actin ACT11 and recognizes subclasses 2 and 3 of vegetative and subclasses 4 and 5 of pollen specific plant actins (arabidopsis ACT1, 3, 4, 7, 11, 12).
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit