Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actin2 Antibody (mAb13a), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253122AF532
Description
Actin2 Monoclonal specifically detects Actin2 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Actin2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Apoptosis, Cancer, Cellular Signaling | |
This mouse monoclonal antibody was generated against plant actin ACT2 and recognizes subclasses 1 and 3 of vegetative plant actins (arabidopsis ACT2, 8, 11). | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
mAb13a | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ACT2, actin 2, Actin-2 | |
Purified Arabidopsis Actin2 | |
0.1 mL | |
Primary | |
Plant | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction