Learn More
Actin8 Mouse anti-Plant, Alexa Fluor™ 532, Clone: mAbGPa, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP253121AF532
Catalog No. NP23121A32
Add to cart
Description
Description
Actin8 Monoclonal antibody specifically detects Actin8 in Plant samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
Actin8
mAbGPa
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Plant
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Purified Arabidopsis actin (ACT8)
0.1 mL
Primary
This mouse monoclonal antibody was generated against plant actin ACT8 and recognizes all tested subclasses of plant actins (arabidopsis ACT1, 2, 3, 4, 7, 8, 11, 12) and Dictyosetelium actin.