Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Actinomycin D, 7-Amino, ≥85%, Calbiochem™,

Click to view available options
Quantity:
1mg
Packaging:
Glass Bottle
7240-37-1
C62H87N13O16
1270.45
MFCD00077295
YXHLJMWYDTXDHS-UHFFFAOYNA-N
7-aad, 7-aminoactinomycin d
131675298
2,7-diamino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C(N)=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O
7240-37-1
C62H87N13O16
1270.45
MFCD00077295
YXHLJMWYDTXDHS-UHFFFAOYNA-N
7-aad, 7-aminoactinomycin d
131675298
2,7-diamino-4,6-dimethyl-3-oxo-N1,N9-bis[2,5,9-trimethyl-1,4,7,11,14-pentaoxo-6,13-bis(propan-2-yl)-hexadecahydro-1H-pyrrolo[2,1-i]1-oxa-4,7,10,13-tetraazacyclohexadecan-10-yl]-3H-phenoxazine-1,9-dicarboxamide
CC(C)C1NC(=O)C(NC(=O)C2=C3N=C4C(OC3=C(C)C(N)=C2)=C(C)C(=O)C(N)=C4C(=O)NC2C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C3CCCN3C(=O)C(NC2=O)C(C)C)C(C)OC(=O)C(C(C)C)N(C)C(=O)CN(C)C(=O)C2CCCN2C1=O

Recommended Storage

−20°C, Stable for up to 2 months at −20°C

Color Purple-Red
Quantity 1mg
Packaging Glass Bottle
Physical Form Solid

Recommended Storage : −20°C, Stable for up to 2 months at −20°C