Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Actinomycin D (Red Crystals), Fisher BioReagents
Click to view available options
Quantity:
5 mg
10 mg
Packaging:
Poly Micro Tube
Description
This antibiotic is a widely used transcription inhibitor.
Specifications
Type | Actinomycin D |
Physical Form | Powder/Solid |
Melting Point | 245°C |
Extinction Coefficient | Report |
Assay Percent Range | ≥98 % |
Merck Index | 15, 2797 |
Packaging | Poly Micro Tube |
Solubility Information | Soluble in Methanol |
Formula Weight | 1255.44 |
Percent Purity | 99.0% min. (HPLC), 98% min. (TLC) |
Show More |
Safety and Handling
TSCA | Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes. |
Recommended Storage | 4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction