Learn More
Action Pump Plastic Siphon Drum Pumps
Choice of polyethylene or polypropylene models
Manufacturer: Action Pump 507
Description
Choice of polyethylene or polypropylene models
- For dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems.
- Maximum delivery rate varies with model
- Use for 15-30 and 55 gal. (57-114, 208L) drums with a 2 in. (5.1cm) bung
- Some units can be used with 5 gal. pail with separate adapter Heavy-duty model available
Specifications
PVC | |
40°F | |
140°F | |
15 to 30 or 55 gal. drum | |
22 oz./stroke | |
1.36 kg |
111.76 cm | |
Drum Pump | |
Dispensing of dilute acids, caustics, oils, waxes, disinfectants without leakage problems | |
44 in. | |
3 lb. |