Ansell ActivArmr® 97-009 Multipurpose Heavy Duty Glove
Versatile protection for all your tough jobs. Rugged ActivArmr Multipurpose Heavy Duty gloves offer the ultimate palm and knuckle protection (Cut Level 4, Abrasion Level 4) with reinforced ridges. Features Ansell's patented Intercept Technology.
Manufacturer: Ansell 97009 L 111816
 View more versions of this product
Catalog No. 19-010-019
  Check Availability
Add to Cart
    Specifications
    Cuff Style Velcro
    Exterior Finish Exterior coated
    For Use With (Application) Demolition, DIY, Handling sheet metal, Masonry, Woodwork, Digging, General construction, Heavy cutting/ sawing, Steel work (tying)
    Length (English) 10-11 in.
    Length (Metric) 24.3 - 28.4cm
    Material Nylon
    Liner Material Lycra, Stainless steel, Kevlar, Nylon
    Size L
    Model 97-009
    Quantity 12
    Coated Area Palm
    Color Black, gray and orange
    Cut Resistance Rating Level 4
    • The rugged ActivArmr Multipurpose Heavy Duty glove is engineered for the most demanding jobs
    • Ansell's patented Intercept Technology, reinforced with stainless steel and Dupont Kevlar, delivers ultimate protection from punctures, cuts and abrasions (Cut Level 4, Abrasion Level 4)
    • Reinforced ridges offer additional palm and knuckle protection
    • A foam nitrile coating provides enhanced wet or dry grip
    • “Second skin” feel gives dexterity and precision all day long
    • The glove features an adjustable wrist closure with pull-tab and ID nameplate
        Compliance
        ANSI Level 4 Abrasion Resistance; ANSI Level 4 Cut Resistance
  • Description & Specifications
Related Products
Product Content Feedback