Learn More
Ansell ActivArmr® 97-009 Multipurpose Heavy Duty Glove
Versatile protection for all your tough jobs. Rugged ActivArmr Multipurpose Heavy Duty gloves offer the ultimate palm and knuckle protection (Cut Level 4, Abrasion Level 4) with reinforced ridges. Features Ansell's patented Intercept Technology.
Manufacturer:  Ansell 97009 M 111815
 View more versions of this product
Catalog No. 19-010-018
  Check Availability
Add to cart
Specifications
Description
  • The rugged ActivArmr Multipurpose Heavy Duty glove is engineered for the most demanding jobs
  • Ansell's patented Intercept Technology, reinforced with stainless steel and Dupont Kevlar, delivers ultimate protection from punctures, cuts and abrasions (Cut Level 4, Abrasion Level 4)
  • Reinforced ridges offer additional palm and knuckle protection
  • A foam nitrile coating provides enhanced wet or dry grip
  • “Second skin” feel gives dexterity and precision all day long
  • The glove features an adjustable wrist closure with pull-tab and ID nameplate
Description & Specifications
Specifications
Palm
Exterior coated
Demolition, DIY, Handling sheet metal, Masonry, Woodwork, Digging, General construction, Heavy cutting/ sawing, Steel work (tying)
Lycra, Stainless steel, Kevlar, Nylon
97-009
Black,Gray,Orange
24.3 - 28.4cm
Level 4
Velcro
Nylon
Medium
12
10-11 in.