Ansell ActivArmr; 97-008 Multipurpose Medium Duty Glove

Versatile protection for all your tough jobs. Rugged ActivArmr Multipurpose Medium Duty gloves are ideal for such projects that involve working with rough materials and power tools. An advanced layer of DuPoint Kevlar provides durability and tough protection against cuts and harsh abrasions (Cut Level 2, Abrasion Level 4).

Manufacturer: Ansell 97008 S 111810

 View more versions of this product

Catalog No. 19-010-021

  Check Availability
Add to Cart

    Specifications

    Length (English) 10-11 in.
    Size S
    Model 97-008
    Coated Area Palm
    Color Black, gray and orange
    Cuff Style Velcro
    Exterior Finish Exterior coated
    For Use With (Application) Cutting, Sanding, Sawing, Piping, Landscaping, General handling, Flooring, Tiling, Roofing, General maintenance
    Length (Metric) 24.3 - 28.4cm
    Material Nylon
    Liner Material Nylon; Spandex; Kevlar
    Quantity 12
    Cut Resistance Rating Level 2

    • The ActivArmr Multipurpose Medium Duty glove is ideal for such projects as working with rough materials and power tools
    • An advanced layer of DuPont Kevlar provides durability and tough protection against cuts and harsh abrasions (Cut Level 2, Abrasion Level 4)
    • Reinforced ridges offer extra palm protection
    • A foam nitrile coating provides enhanced wet or dry grip
    • “Second skin” feel gives dexterity and precision all day long
    • The glove features an adjustable wrist closure with pull-tab and ID nameplate

    Compliance

    ANSI Level 4 Abrasion Resistance; ANSI Level 2 Cut Resistance

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit