Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell ActivArmr™ 97-125 Cut and Impact Protection Gloves
Click to view available options
Size:
8
9
10
11
12
Description
- HIVIZ™ design increases awareness of users for safety
- Double-dip nitrile coating provides grip and control when working with oils and other liquids
- HPPE seamless knit glove keeps hands cool and comfortable
- Double-dip nitrile coating defends against oil exposure and abrasion injuries
- Designed for dynamic environments which have impact and cut hazards present
Specifications
ANSI Abrasion Rating | ANSI 5 |
Certifications/Compliance (Europe) | CE, EN 420:2003 + A1:2009; Category II |
Construction | Knit |
Coating Material | Nitrile |
Color | Gray, Hi-vis Orange |
Cut Resistance Rating | ANSI A4 |
Product Type | Cut-resistant Gloves |
Size | 8 |
EN388 Standard | EN 388:4X43CP |
For Use With (Application) | Engagement and release of drill string slips, drilling assembly and completion, handling jagged metal, production Services, drilling/extraction, heavy equipment servicing, transport and logistics |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction