Learn More
Ansell™ ActivArmr™ Two-Layer Gloves
Ansell™ ActivArmr™ Two-Layer Gloves feature a unique combination of warmth, dexterity, and flexibility, allowing the user feel everything but the cold
Manufacturer:  Ansell™ 97631060
 View more versions of this product
Catalog No. 19-500-761
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • PVC coating provides excellent grip and flexibility, even in the coldest temperatures
  • Flexible nylon exterior protects against mechanical risks, including cut, and contours to the hand permitting maximum dexterity
  • Soft and insulative acrylic interior provides comfort and warmth
Specifications
Specifications
EN420:2003 + A1:2009; Category ll; REACH Compliant; EN51112X
Construction; Outdoor Winter Use; Cold Storage; Utlities
250 to 285 mm
6
Safety
Black
9.84 to 11.22 in.
Acrylic, Nylon, PVC
Glove