Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activated charcoal, iodinated, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
Description
Filter aid
Specifications
Quantity | 250g |
Color | Black |
Packaging | Glass bottle |
Merck Index | 14, 1807 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 12 |
Physical Form | Thin Rods |
Percent Purity | 5%, exact strength on the certificate of analysis |
Chemical Name or Material | Activated charcoal |
Safety and Handling
GHS H Statement
Flammable solid.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 231-153-3