Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activated Charcoal, Powder, USP, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7440-44-0
C
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
[C]
7440-44-0
C
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
[C]

CA191, 7440-44-0, C

Spectrum™ Activated Charcoal, Powder, USP is carbon that has been treated to create low-volume pores that increase the area available for chemical reactions and adsorption. The most common pharmaceutical uses of activated charcoal is as a purification agent and antitoxin. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 15%
Ignition Residue 4%
Packaging Poly Pail
Quantity 2.5 kg
CAS Max % 100%