Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activated Charcoal, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
7440-44-0
C
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
[C]
7440-44-0
C
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
[C]

C1955, 7440-44-0, C

Spectrum™ Activated Charcoal is a form of carbon processed with steam or chemicals to generate many low-volume pores that increases the surface area available for absorption or chemical reactions. Because of its huge degree of micro-porosity, one gram can have up to 1000 m2 of surface area. Activated charcoal has many uses including gas purification, decaffeination, gold purification, metal extraction, water purification, medicine, sewage treatment, air filters, gas masks, and respirators along with filters in compressed air tanks. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Ignition Residue 4%
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 100%