Learn More
Enzo Life SciencesSupplier Diversity Partner Src (human), (recombinant) (GST-tag) (5 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADIPPK460Z
Alternative name: CSK. LCK. MW: ~85kDa. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-hydrogen chloride, pH 7.5, containing 150mM sodium chloride, 0.25mM DTT, 0.1mM EGTA, and 30% glycerol. Source: Produced in Sf9 insect cells using an N-terminal GST-tag. UniProt: P12931. Application: Kinase Assay, Long Term Storage: -80°C
Catalog No. 50-667-375