Learn More
Activin A Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0074-7L18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212009AF532
Catalog No. NP21209A532
Add to cart
Description
Description
Activin A Monoclonal antibody specifically detects Activin A in Human samples. It is validated for Western Blot.
Specifications
Specifications
Activin A
MM0074-7L18
Western Blot
activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
INHBA
Human Activin A
0.1 mL
Primary
3624