Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin A Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0074-7L18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212009AF594

Catalog No. NP21209A594


Add to cart

Description

Description

Activin A Monoclonal antibody specifically detects Activin A in Human samples. It is validated for Western Blot.
Specifications

Specifications

Activin A
MM0074-7L18
Western Blot
Conjugated, Purified
INHBA
Human Activin A
Protein G purified
Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Stem Cell Signaling Pathway
Primary
3624.0
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
activin AB alpha polypeptide, Activin beta-A chain, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FSH-releasing protein, inhibin beta A chain, inhibin beta A subunit, Inhibin, beta-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit