Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Activin B/Inhibin beta B Mouse anti-Human, Clone: MM0075-9M29, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | Activin B/Inhibin beta B |
---|---|
Clone | MM0075-9M29 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin), Blocking Assay |
Classification | Monoclonal |
Description
Activin B/Inhibin beta B Monoclonal specifically detects Activin B/Inhibin beta B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
Activin B/Inhibin beta B | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human | |
activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 | |
INHBB | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0075-9M29 | |
Western Blot, Immunohistochemistry (Paraffin), Blocking Assay | |
Unconjugated | |
Mouse | |
Cytokine Research, Stem Cell Signaling Pathway | |
Q53T31 | |
3625 | |
IgG1 | |
Protein G purified |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title