Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activin RIIA Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI5D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270079AF700

Catalog No. N270079A700


Add to cart

Description

Description

Activin RIIA Monoclonal antibody specifically detects Activin RIIA in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Activin RIIA
OTI5D1
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ACVR2A
Recombinant protein fragment corresponding to amino acids 214-513 of human Activin RIIA (NP_001607) produced in E.coli.
Immunogen affinity purified
Cancer
Primary
92.0
Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
activin A receptor, type II, activin A receptor, type IIA, Activin receptor type IIA, activin receptor type-2A, ACTRII, ACTRIIA, ACVR2ACTR-IIA, EC 2.7.11, EC 2.7.11.30
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit