Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gatorade Activity Drinks Liquid Concentrates

Package yields 6 gallons

$67.75

Specifications

Format Concentrate
Product Type Activity Drinks
Quantity 4/case
Volume (English) 1 gal
View More Specs

Products 2
Catalog Number Mfr. No. Flavor Price Quantity & Availability  
Catalog Number Mfr. No. Flavor Price Quantity & Availability  
19-313-500
View Documents
Gatorade
30803984
Lemon-Lime
Case of 4 for $67.75
Add to cart
 
19-313-499
View Documents
Gatorade
30833305
Fierce Grape
Case of 4 for $67.75
Add to cart
 
Specifications

Specifications

Concentrate
Activity Drinks
4/case
1 gal
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit