Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACTL9 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548851
Description

Specifications
ACTL9 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8TC94 | |
ACTL9 | |
KLH conjugated synthetic peptide between 23-50 amino acids from the N-terminal region of human ACTL9. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ACTL9 | |
Actin-like protein 9, ACTL9, ACTL7C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284382 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction