Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ACTR3 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Supplier: Novus Biologicals™ NBP2518460.5MG
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-418 of Human ACTR3 The Recombinant Human ACTR3 Protein is derived from E. coli. The Recombinant Human ACTR3 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol | |
TMW: 49.8kDa | |
0.5mg | |
actin-related protein 3, ARP3 (actin-related protein 3, yeast) homolog, ARP3 actin-related protein 3 homolog (yeast), ARP3Actin-like protein 3 | |
Unlabeled |
SDS-Page | |
10096 | |
>85%, by SDS-PAGE | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
ACTR3 | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis