Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACTR6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109359
Description
Immunogen sequence: VLPDFSTIKK GFCKPREEMV LSGKYKSGEQ ILRLANERFA VPEILFNPSD IGIQEMGIPE AIVYSIQNLP EEMQPHFFKN IVLTGGNSLF PGFRDRVYSE VRCLTPTDYD VSVVLPENPI TYAWEGGKLI SENDDFEDMV VTREDYEENG HSVCEEKFDI.
The function of this protein remains unknown.
Specifications
ACTR6 | |
Polyclonal | |
Unconjugated | |
ACTR6 | |
2010200J04Rik; actin related protein 6; actin-related protein 6; actin-related protein 6-like protein; Actr6; Arp6; ARP6 actin related protein 6 homolog; ARP6 actin-related protein 6; ARP6 actin-related protein 6 homolog; ARP6 actin-related protein 6 homolog (yeast); ArpX; AU044443; CDA12; FLJ13433; h hARPX; hArp6; hARPX; HSPC281; LOW QUALITY PROTEIN: actin-related protein 6; mArp6; MSTP136 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
314718, 64431, 67019 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.73 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9D864, Q9GZN1 | |
ACTR6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 237-396 of human ACTR6 (NP_071941.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction