Learn More
Sekisui Diagnostics™ Acucy™ System, Bundle Purchase
The Acucy™ System is composed of the Acucy Reader, Acucy Printer, and other accessories required to start running Acucy tests (includes 4 test kits).
Manufacturer:  Sekisui Diagnostics™ 1039B
Catalog No. 22-321-127
Add to cart
This item is not returnable. View return policy
Description
Description
  • Flex Purchase is a discounted Acucy System that requires a purchase commitment of at least 8 Acucy Influenza A&B kits
  • Innovative Composite Comparator with molecular and culture
  • Definitive and standardized results
  • Available connectivity
  • Read Now or Walk Away mode
  • Results within 15 minutes
  • CLIA-waived procedure
  • QC Inside – removes added expense of performing quality control testing
Specifications
Specifications
Nasal and Nasopharyngeal Swabs
Acucy Reader, Acucy Printer, and other accessories required to start running Acucy tests