Learn More
DWK Life Sciences Acura™ Self-Refilling Microdispenser Pipette
Three sizes of pipettes for Acura™ manual 865 Self Refilling Microdispenser Pipette.
Supplier: DWK Life Sciences W835001
Description
- Excellent ergonomics
- Soft plunger activation
- Versatile feeding through bottle, tubing or syringe
- No need for consumables
- Swift-set in-lab calibration
- Autoclavable fully assembled at 121
Specifications
Autoclavable | |
1 | |
Self-Refilling Microdispensor Pipette | |
1 Each |
Acura™ 865 | |
Acura™ | |
5 to 50 μL |