Learn More
DWK Life Sciences Acura™ Self-Refilling Microdispenser Pipette
Three sizes of pipettes for Acura™ manual 865 Self Refilling Microdispenser Pipette.
$586.44 - $626.78
Specifications
Number of Channels 1
Autoclavable Autoclavable
Model Acura™ 865
Type Self-Refilling Microdispensor Pipette
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
13-709-550 View Documents DWK Life Sciences
W835001
5 to 50 μL Each for $623.67
Add to cart
13-709-551 View Documents DWK Life Sciences
W835002
20 to 200 μL Each for $586.44
Add to cart
13-709-552 View Documents DWK Life Sciences
W835004
100 to 1000 μL Each for $626.78
Add to cart
Description
Description
  • Excellent ergonomics
  • Soft plunger activation
  • Versatile feeding through bottle, tubing or syringe
  • No need for consumables
  • Swift-set in-lab calibration
  • Autoclavable fully assembled at 121
Specifications
Specifications
1
Autoclavable
Acura™ 865
Self-Refilling Microdispensor Pipette