Learn More
Abnova™ ACVR2B 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000093T02
Catalog No. 89-932-509
Add to cart
Description
Description
Specifications
Specifications
293T
BC096245.1
pCMV-ACVR2B full-length
100 μL
Human
ACVR2B 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 34.2kDa
Transient overexpression cell lysate was tested with anti-ACVR2B antibody (Mfr. No. H00000093-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot