Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACY3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113216
Description
ACY3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
ACY3 belongs to the aspA/astE family. Aspartoacylase subfamily. Catalytic activity: N-acyl-L-aspartate + H(2)O = a carboxylate + L-aspartate.
Specifications
ACY3 | |
Polyclonal | |
Unconjugated | |
Acy3 | |
0610006H10Rik; AA3; AAIII; Acy3; ACY-3; Acylase III; aminoacylase 3; Aminoacylase III; Aminoacylase-3; Aspa2; aspartoacylase (aminoacylase) 3; aspartoacylase (aminocyclase) 3; aspartoacylase 3; aspartoacylase-2; aspartoacylase-3; AW107362; EGK_05921; HCBP1; HCV core-binding protein 1; hepatitis C virus core-binding protein 1; LOW QUALITY PROTEIN: N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming); RGD1305394 | |
Rabbit | |
Protein G | |
RUO | |
91703 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96HD9 | |
Acy3 | |
Recombinant Human N-acyl-aromatic-L-amino acid amidohydrolase (carboxylate-forming) protein (5-319AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction