Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acyclovir, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
100 mg
1 g
5 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail
59277-89-3
C8H11N5O3
225.21
MKUXAQIIEYXACX-UHFFFAOYSA-N
2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-3H-purin-6-one
NC1=NC(=O)C2=C(N1)N(COCCO)C=N2
59277-89-3
C8H11N5O3
225.21
MKUXAQIIEYXACX-UHFFFAOYSA-N
2-amino-9-[(2-hydroxyethoxy)methyl]-6,9-dihydro-3H-purin-6-one
NC1=NC(=O)C2=C(N1)N(COCCO)C=N2

A1816, 59277-89-3, C8H11N5O3

Spectrum™ Acyclovir, USP is the most commonly used antiviral drug and is on the World Health Organization′s list of essential medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 g
CAS Max % 1
Percent Purity 98 to 101%