Learn More
BioVision IncSupplier Diversity Partner Adenosine Deaminase Activity Assay Kit (Colorimetric): Adenosine Deaminase Activity Assay Kit (Colorimetric): Simple & convenient assay to measure ada activity in a variety of samples. Limit of quantification: 1 mU recombinant Adenosine. 100 assay DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K321100
Adenosine Deaminase Activity Assay Kit (Colorimetric): Adenosine Deaminase (ADA) (E.C. 3.5.4.4.) is an enzyme that catalyzes the conversion of adenosine and 2¿-deoxyadenosine to inosine and 2¿-deoxyinosine. Adenosine Deaminase is widely distributed in various tissues and cells. It is present in the cytosol, nucleus and found associated with dipeptidyl peptidase-4 on the cell membrane. ADA2 was first found in the spleen but is predominantly found in the plasma and serum. Increased serum ADA levels are found in certain infectious diseases such as tuberculosis and various liver diseases such as acute hepatitis, alcoholic hepatic fibrosis, chronic active hepatitis to name a few. In BioVision's ADA Activity Assay, inosine formed from the breakdown of adenosine is converted to uric acid with ADA Convertor and ADA Developer. The uric acid is measured at OD 293 nm. The kit measures total Activity of Adenosine Deaminase with limit of quantification of 1 mU recombinant Adenosine Deaminase.
Catalog No. 50-111-9691