Learn More
Fisherbrand™ ADA Height Wood Fixed Height Table, 48 in. Wide, Black Phenolic Work Surface
Fisherbrand™ Wood Fixed Height Tables bring quality, warmth and durability to your laboratory.
Description
- Made from hand selected premium rotary cut maple or rift cut oak veneers.
- Robotically applied environmentally friendly finishes are chemically resistant.
- Wood Fixed Height Tables are furnished with 4.875 in. tall apron rails.
- The legs have a 2.25 x 2.25 in. construction and inculdes leveling feet and a black trim boot at the base of the leg.
Compliance
SEFA 8
Specifications
Fixed Height Table | |
Honey Stain | |
33.5 in. | |
2 years | |
76.2 cm | |
Maple Veneer |
Table Frame, Phenolic Work Surface, Leveling Feet and Black Trim Boot | |
30 in. | |
85.1 cm | |
121.9 cm | |
48 in. |