Learn More
Fisherbrand™ ADA Height Wood Panel End Supports
Fisherbrand™ Wood Panel End Supports bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB9200L32M6
Add to cart
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Wood Panel End Supports are used in place of a cabinet at one end of a cabinet run to support drawer frames or apron rails.
  • Can be used to support the intersection of two aprons or rails.
  • Each panel has 2 leveling feet
Compliance
SEFA 8
Specifications
Specifications
Chili Stain
57.2 cm
82.6 cm
1 in.
22.5 in.
32.5 in.
Wood (Maple Veneer)
2.5 cm